Diethylether 99+% stabilised, pure
Metal drum, 25 L
Leverandør: Thermo Fisher Scientific
Diethylether 99+% stabilised, pure
Katalog # ACRO123990250
Leverandør: Thermo Fisher Scientific
CAS nummer: 60-29-7
Gå til:
Product Details & Documents
Synonymer: Ether
- CAS nummer:60-29-7
- Formel:(CH₃CH₂)₂O
- MDL nummer:MFCD00011646
- Molecular Weight:74.12 g/mol
- Merck Index:12,03852
- Kogende Pt:34,6 °C (760 mmHg)
- Smeltende Pt:–116 °C
- Tæthed:0,714 g/cm³
- Flash Pt:–45 °C
- FN:1155
- ADR:3,I
- :stabiliseret med BHT/Ionol
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-467-2
- NÅ:01-2119535785-29
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3510 to 1.3540 (20°C, 589 nm)
- Water:≤0.2 % (Coulometric)
- GC:≥99.0 %
- Color scale:≤10 APHA
- Stabilizer:5 to 10 ppm BHT
- Residue after evaporation:≤0.002 %
- Peroxides:≤0.001 %
Specifikationer
- Pakning:Metal drum
- Pakn:25 L



