Iodmethan 99% stabilised
Glass bottle, 2,5 kg
Leverandør: Thermo Fisher Scientific
Iodmethan 99% stabilised
Katalog # ACRO122370025
Leverandør: Thermo Fisher Scientific
CAS nummer: 74-88-4
Gå til:
Product Details & Documents
Synonymer: Methyliodid
- CAS nummer:74-88-4
- Formel:CH₃I
- MDL nummer:MFCD00001073
- Molecular Weight:141.94 g/mol
- Merck Index:12,06161
- Kogende Pt:42,5 °C (760 mmHg)
- Smeltende Pt:–66 °C
- Tæthed:2,28 g/cm³
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-819-5
- FN:2644
- ADR:6.1,I
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.5290 to 1.5310 (20°C, 589 nm)
- Water:≤0.1 %
- GC:≥98.5 %
- Color scale:≤150 APHA
- Stabilizer:Copper
- Non-volatile matter:≤0.01 %
- Heavy metals (as Pb):≤0.002 %
- Hydrolyzable substance:Passes test
Specifikationer
- Pakning:Glass bottle
- Pakn:2,5 kg



