Iodacetamid 98%
Plastic bottle, 500 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: 2-Iodoacetamide
- CAS nummer:144-48-9
- Formel:ICH₂CONH₂
- MDL nummer:MFCD00008028
- Molecular Weight:184.96 g/mol
- Kogende Pt:266 °C (1013 hPa)
- Smeltende Pt:>90 °C
- Opbevaringstemperatur:Køleskab
- EINECS:205-630-1
- FN:2923
- ADR:8,II
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White to yellow to beige Powder or crystals
- Infrared spectrum:Conforms
- Melting point:≥90 °C
- Titration Argentometric:≥97.5 % (ex I)
Specifikationer
- Pakning:Plastic bottle
- Pakn:500 g



