8-Quinolinol ACS
Plastic bottle, 2,5 kg
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: 8-Hydroxyquinoline
- CAS nummer:148-24-3
- Formel:C₉H₇NO
- MDL nummer:MFCD00006807
- Molecular Weight:145.16 g/mol
- Merck Index:13,04869
- Kogende Pt:267 °C (752 mmHg)
- Smeltende Pt:72…74 °C
- Tæthed:1,03 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:205-711-1
- FN:2811
- ADR:6.1,III
Documents
ACRO12190
SDS
Specifikationer
Specifikationstestresultater
- Appearance:White to light yellow to light beige Crystalline powder or crystals or flakes
- Infrared spectrum:Conforms
- Melting point:72.5 to 74.0 °C
- Residue after ignition:≤0.05 %
- Suitability:(for Mg determination) Passes test
- Sulfate (SO4):≤0.02 %
- Insoluble matter:≤0.05 % (in alcohol)
Specifikationer
- Pakning:Plastic bottle
- Pakn:2,5 kg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



