trans-4-Hydroxy-L(-)-prolin 99+%
Glass bottle, 500 g
Leverandør: Thermo Fisher Scientific
trans-4-Hydroxy-L(-)-prolin 99+%
Katalog # ACRO121785000
Leverandør: Thermo Fisher Scientific
CAS nummer: 51-35-4
Gå til:
Product Details & Documents
Synonymer: L-4-Hydroxyproline, (S)-(-)-trans-4-Hydroxyproline, L(-)-trans-4-Hydroxyproline, 4-Hydroxy-L-Proline, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, H-Hyp-OH
- CAS nummer:51-35-4
- Formel:C₅H₉NO₃
- MDL nummer:MFCD00064320
- Molecular Weight:131.13 g/mol
- Merck Index:13,04866
- Kogende Pt:296 °C (1013 hPa)
- Smeltende Pt:274…275 °C (dec.)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-091-9
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White Crystalline powder or crystals
- Infrared spectrum:Conforms
- Loss on drying:≤0.2 % (105°C, 3 hrs)
- Sulfated ash:≤0.1 %
- UV transmittance:≥95 % (c=10, H2O, 430 nm) 1 cm cell
- Specific optical rotation:-74° to -77° (20°C, 589 nm) (c=5, H2O)
- Titration with HClO4:99.0 to 101.0 %
- Chloride (Cl):≤200 ppm
- Sulfate (SO4):≤200 ppm
- Ammonium (NH4):≤200 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤10 ppm
- Other amino acids:≤0.5 % (TLC)
Specifikationer
- Pakning:Glass bottle
- Pakn:500 g



