Anisol 99%, pure
Glass bottle, 2,5 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Methyl phenyl ether, Methoxybenzene
- CAS Number:100-66-3
- Formula:C₇H₈O
- MDL Number:MFCD00008354
- Molecular Weight:108.14 g/mol
- Merck Index:13,00672
- Boiling Pt:154 °C (760 mmHg)
- Melting Pt:–37 °C
- Density:0,99 g/cm³
- Flash Pt:45 °C
- Storage Temperature:Stue temperatur
- EINECS:202-876-1
- UN:2222
- ADR:3,III
Documents
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5160 to 1.5190 (20°C, 589 nm)
- Water:≤0.1 % (K.F.)
- GC:≥98.5 %
Specifications
- Pakning:Glass bottle
- Pakn:2,5 L



