1,2,4-Triazol 99.5%
Plastic bottle, 500 g
Leverandør: Thermo Fisher Scientific
1,2,4-Triazol 99.5%
Katalog # ACRO139285000
Leverandør: Thermo Fisher Scientific
CAS nummer: 288-88-0
Gå til:
Product Details & Documents
Synonymer: 1,2,4-1H-Triazole, 1H-1,2,4-Triazole
- CAS nummer:288-88-0
- Formel:C₂H₃N₃
- MDL nummer:MFCD00005228
- Molecular Weight:69.07 g/mol
- Merck Index:12,09735
- Kogende Pt:260 °C (760 mmHg)
- Smeltende Pt:119…122 °C
- Tæthed:1,394 g/cm³ (25 °C)
- Flash Pt:140 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:206-022-9
Documents
ACRO13928
SDS
Specifikationer
Specifikationstestresultater
- Appearance:White Powder and/or flakes
- Infrared spectrum:Conforms
- Melting point:119 to 122 °C
- Water:≤0.2 % (K.F.)
- HPLC:≥99.4 %
Specifikationer
- Pakning:Plastic bottle
- Pakn:500 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



