Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Cyclopentanon 99+%, pure

Cyclopentanon 99+%, pure

Plastic drum, 10 L
Catalog # ACRO111530100
Irritant
Flammable
Cyclopentanon 99+%, pure
Catalog # ACRO111530100
CAS Number:  120-92-3
Cyclopentanon 99+%, pure
Cyclopentanon 99+%, pure

Plastic drum, 10 L

CAS Number:  120-92-3
Price
5.970,00 kr
Log in for availability
1 * 10 L
Product Details & Documents
  • CAS Number:120-92-3
  • Formula:C₅H₈O
  • MDL Number:MFCD00001409
  • Molecular Weight:84.12 g/mol
  • Merck Index:13,02771
  • Boiling Pt:130…131 °C (760 mmHg)
  • Melting Pt:–51 °C
  • Density:0,95 g/cm³
  • Flash Pt:26 °C
  • Storage Temperature:Stue temperatur
  • EINECS:204-435-9
  • UN:2245
  • ADR:3,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4360 to 1.4380 (20°C, 589 nm)
  • GC:
    ≥99.0 %
  • Stabilizer:
    0.01 % alpha-Tocopherol

Specifications

  • Pakning:
    Plastic drum
  • Pakn:
    10 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.