Cyclopentanon 99+%, pure
Glass bottle, 2,5 L
Leverandør: Thermo Fisher Scientific
Cyclopentanon 99+%, pure
Katalog # ACRO111530025
Leverandør: Thermo Fisher Scientific
CAS nummer: 120-92-3
Gå til:
Product Details & Documents
- CAS nummer:120-92-3
- Formel:C₅H₈O
- MDL nummer:MFCD00001409
- Molecular Weight:84.12 g/mol
- Merck Index:13,02771
- Kogende Pt:130…131 °C (760 mmHg)
- Smeltende Pt:–51 °C
- Tæthed:0,95 g/cm³
- Flash Pt:26 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:204-435-9
- FN:2245
- ADR:3,III
Documents
ACRO11153
SDS
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4360 to 1.4380 (20°C, 589 nm)
- GC:≥99.0 %
- Stabilizer:0.01 % alpha-Tocopherol
Specifikationer
- Pakning:Glass bottle
- Pakn:2,5 L
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



