o-Cresol 99%
Plastic drum, 10 kg
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: o-Methylphenol
- CAS nummer:95-48-7
- Formel:C₇H₈O
- MDL nummer:MFCD00002226
- Molecular Weight:108.14 g/mol
- Merck Index:13,02604
- Kogende Pt:191 °C (760 mmHg)
- Smeltende Pt:30…32 °C
- Tæthed:1,04 g/cm³
- Flash Pt:81 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:202-423-8
- FN:3455
- ADR:6.1,II
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Colorless to amber Low melting solid or liquid (May darken on storage)
- Infrared spectrum:Conforms
- Water:≤0.2 % (K.F.)
- GC:≥98.5 %
Specifikationer
- Pakning:Plastic drum
- Pakn:10 kg



