(2-Hydroxyethyl)trimethylammoniumchlorid 99%
Plastic bottle, 50 g
: 2-hydroxy-N,N,N-trimethylethanaminium chloride
- :67-48-1
- :HOH₂CCH₂N(CH₃)₃Cl
- :MFCD00011721
- :139.62 g/mol
- :13,02226
- :298…304 °C (dec.)
- :~ 1,205 g/cm³ (20 °C)
- :Stue temperatur
- :200-655-4
- Appearance:White Adhering crystals
- Infrared spectrum:Conforms
- Titration Argentometric:98.5 to 101.5 % (on dry substance)
- Water:≤2 %
- Pakning:Plastic bottle
- Pakn:50 g



