(2-Hydroxyethyl)trimethylammoniumchlorid 99%
Plastic drum, 5 kg
Leverandør: Thermo Fisher Scientific
(2-Hydroxyethyl)trimethylammoniumchlorid 99%
Katalog # ACRO110290050
Leverandør: Thermo Fisher Scientific
CAS nummer: 67-48-1
Gå til:
Product Details & Documents
Synonymer: 2-hydroxy-N,N,N-trimethylethanaminium chloride
- CAS nummer:67-48-1
- Formel:HOH₂CCH₂N(CH₃)₃Cl
- MDL nummer:MFCD00011721
- Molecular Weight:139.62 g/mol
- Merck Index:13,02226
- Smeltende Pt:298…304 °C (dec.)
- Tæthed:~ 1,205 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-655-4
Specifikationer
Specifikationstestresultater
- Appearance:White Adhering crystals
- Infrared spectrum:Conforms
- Titration Argentometric:98.5 to 101.5 % (on dry substance)
- Water:≤2 %
Specifikationer
- Pakning:Plastic drum
- Pakn:5 kg



