Phthalsyre 99%
Plastic bottle, 2,5 kg
: Pathalic acid
- :88-99-3
- :C₈H₆O₄
- :MFCD00002467
- :166.13 g/mol
- :12,07527
- :315 - 317 °C (1013 hPa)
- :200…205 °C (dec.)
- :1,576 g/cm³ (20 °C)
- :168 °C
- :Stue temperatur
- :201-873-2
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:200 to 205 °C (Dec)
- Titration with NaOH:≥98.5 %
- Chloride (Cl):≤50 ppm
- Sulfate (SO4):≤50 ppm
- Iron (Fe):≤50 ppm
- Heavy metals (as Pb):≤10 ppm
- Pakning:Plastic bottle
- Pakn:2,5 kg



