Majsolie
Plastic bottle, 2,5 kg
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
- CAS nummer:8001-30-7
- MDL nummer:MFCD00130865
- Smeltende Pt:-10 °C
- Tæthed:0,92 g/cm³
- Flash Pt:>200 °C
- Opbevaringstemperatur:Stue temperatur
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Clear yellow Oily liquid
- Infrared spectrum:Conforms
- Refractive index:1.4730 to 1.4760 (20°C, 589 nm)
- Acid value:≤0.5 mg KOH/g
- Peroxide value:≤5 meq O2/kg
Specifikationer
- Pakning:Plastic bottle
- Pakn:2,5 kg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



