Galdesyre 97%
Glass bottle, 500 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: 3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid, 3α,7α,12α-Trihydroxycholan-24-oic acid, (R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-Trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic acid, 3α,7α,12α-Trihydroxycholanic acid, 3α,7α,12α-Trihydroxy-5β-cholanic acid
- CAS nummer:81-25-4
- Formel:C₂₄H₄₀O₅
- MDL nummer:MFCD00003672
- Molecular Weight:408.58 g/mol
- Merck Index:13,02223
- Kogende Pt:545 °C (1013 hPa)
- Smeltende Pt:197…202 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:201-337-8
Specifikationer
Specifikationstestresultater
- Appearance:White to light beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:197 to 202 °C
- Loss on drying:≤1 % (135 °C)
- Heavy metals:≤20 ppm
- Sulfated ash:≤0.1 %
- TLC:≥96.0 %
- Specific optical rotation:(20 °C, 589 nm) (c=2, ethanol) > +35°
- Titration with NaOH:≥96.0 %
- UV:at 370 nm A: ≤0.004
- Color scale:≤10 APHA
- Residue after evaporation:≤0.0005 %
- Chloride (Cl):≤0.0005 %
- Sulfate (SO4):≤0.0005 %
- Cadmium (Cd):≤0.000005 %
- Copper (Cu):≤0.000002 %
- Ammonia (NH3):≤0.002 %
Specifikationer
- Pakning:Glass bottle
- Pakn:500 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



