VWR®, Overalls, SPP
hvid, XL
Katalog # 113-8313
Leverandør: VWR Collection
Gå til:
Product Details & Documents
Breathable overalls made of spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
- With collar
- Two-way nylon zip
- Elasticated cuffs and ankles
Weight: 40 g/m².
Colour: White.
Specifikationer
- Tøj størrelse:XL
- Farve:hvid
- Seq:03
- Cat. No.:113-8313
- Vægt:40 g/m²
- Model:With collar
- Outfit applikationer:General Purpose|Industrial