VWR®, Overalls, SPP
hvid, L
113-8312
Breathable overalls made of spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
- With collar
- Two-way nylon zip
- Elasticated cuffs and ankles
Weight: 40 g/m².
Colour: White.
- Tøj størrelse:L
- Farve:hvid
- Seq:02
- Cat. No.:113-8312
- Vægt:40 g/m²
- Model:With collar
- Outfit applikationer:General Purpose|Industrial