VWR®, Overalls, SPP
hvid, M
Catalog # 113-8311
Supplier: VWR Collection
Jump to:
Product Details & Documents
Breathable overalls made of spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
- With collar
- Two-way nylon zip
- Elasticated cuffs and ankles
Weight: 40 g/m².
Colour: White.
Specifications
- Tøj størrelse:M
- Farve:hvid
- Seq:01
- Cat. No.:113-8311
- Vægt:40 g/m²
- Model:With collar
- Outfit applikationer:General Purpose|Industrial