Orders LinkContactUsLinkComponent
Milk Samplers
Milk Samplers
Milk sampler, 100 ml
Katalog # GERB3011
Leverandør:   GERBER FUNKE
Milk Samplers
Katalog # GERB3011
Leverandør:  GERBER FUNKE
Milk Samplers
Milk sampler, 100 ml
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
It is made up of stainless steel with valve for automatic drainage.
Specifikationer
  • Kapacitet:
    100 ml
  • Beskrivelse:
    Milk sampler