Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Phosphate-buffered saline (PBS), Corning®

Phosphate-buffered saline (PBS), Corning®

Phosphate-buffered saline, 1X without calcium and magnesium, 12
Katalog # 21-040-CMX12
Leverandør:   Corning
Phosphate-buffered saline (PBS), Corning®
Katalog # 21-040-CMX12
Leverandør:  Corning
CAS nummer:  
Phosphate-buffered saline (PBS), Corning®
Phosphate-buffered saline (PBS), Corning®

Phosphate-buffered saline, 1X without calcium and magnesium, 12

CAS nummer:  
Price
2.360,00 kr
Log ind for tilgængelighed
1 * 12 Items
Product Details & Documents

Phosphate buffered saline (PBS) is a buffer solution containing sodium chloride, sodium phosphate, and in some formulations potassium chloride and potassium phosphate

  • Low endotoxin - <0.25 EU/ml
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment

PBS helps to maintain a constant pH and is isotonic and non-toxic to cells.

Specifikationer

Specifikationer

  • Pakn:
    12
  • Beskrivelse:
    Phosphate-buffered saline, 1X without calcium and magnesium
  • Cat. No.:
    21-040-CMX12
  • Endotoksinindhold:
    < 0.25 EU/mL
  • Shelf life:
    24 months
  • Mycoplasma:
    Tested Negative
  • Opbevaringstemperatur:
    2...30 °C

Specifikationer