Leather gloves, A271
L, lysebrun
Katalog # PRTWA271TARL
Leverandør: Portwest
Gå til:
Product Details & Documents
Premium quality leather drivers glove with Insulatex™ lining for added warmth and comfort.
- Made from cow grain leather
- Provide mechanical and cold protection
Certifications: EN 388:2016 (3142X), EN 511 (22X)
Specifikationer
- Handske størrelse:L
- Farve:lysebrun
- Seq:01
- Cat. No.:PRTWA271TARL
- Foringsmateriale:Insulatex™
- Ambidextrous/Hånd-specifik:Hånd-specifik
- Model:A271
- EN norm:EN 388:2016 (3142X), EN 511 (22X)
- Engangs/Genbrugelig:Genanvendelig