Beakers, with handle, PP
5000 ml, 257 mm, 203 mm, 100 ml
Katalog # BURK7210-5000
Leverandør: BURKLE
Gå til:
Product Details & Documents
PP, highly transparent.
- With embossed graduation printed in blue
- Ergonomic open or close handle for secure grip
- Functional spout to minimize spills
Ideal for industrial applications.
Specifikationer
- Udv.Ø:203 mm
- Inddelings-interval:100 ml
- Kapacitet:5000 ml
- Højde:257 mm
- Seq:09