Orders LinkContactUsLinkComponent
Beakers, with handle, PP
Beakers, with handle, PP
500 ml, 135 mm, 90 mm, 10 ml
Catalog # BURK7210-0500
Supplier:   BURKLE
Beakers, with handle, PP
Catalog # BURK7210-0500
Supplier:  BURKLE
Beakers, with handle, PP
Beakers, with handle, PP
500 ml, 135 mm, 90 mm, 10 ml
Price
41,90 kr
Log in for availability
1 * 1 Items
Product Details & Documents
PP, highly transparent.
  • With embossed graduation printed in blue
  • Ergonomic open or close handle for secure grip
  • Functional spout to minimize spills
Ideal for industrial applications.
Specifications
  • Udv.Ø:
    90 mm
  • Inddelings-interval:
    10 ml
  • Kapacitet:
    500 ml
  • Højde:
    135 mm
  • Seq:
    06