Accessories for centrifuges
Adapter for 250 ml Corning conical bottle, 200 ml Nunc™ bottle or 175 ml Nalgene conical bottle, 4 x 250 ml (62×145 mm), for TX-1000 rotor
Katalog # 521-3415
Leverandør: Thermo Scientific
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Adapter for 250 ml Corning conical bottle, 200 ml Nunc™ bottle or 175 ml Nalgene conical bottle, 4 x 250 ml (62×145 mm), for TX-1000 rotor
- Seq:04