Orders LinkContactUsLinkComponent
Accessories for centrifuges
Accessories for centrifuges
Adapter for 250 ml Corning conical bottle, 200 ml Nunc™ bottle or 175 ml Nalgene conical bottle, 4 x 250 ml (62×145 mm), for TX-1000 rotor
Katalog # 521-3415
Leverandør:   Thermo Scientific
Accessories for centrifuges
Katalog # 521-3415
Leverandør:  Thermo Scientific
Accessories for centrifuges
Accessories for centrifuges
Adapter for 250 ml Corning conical bottle, 200 ml Nunc™ bottle or 175 ml Nalgene conical bottle, 4 x 250 ml (62×145 mm), for TX-1000 rotor
Price
8.490,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Adapter for 250 ml Corning conical bottle, 200 ml Nunc™ bottle or 175 ml Nalgene conical bottle, 4 x 250 ml (62×145 mm), for TX-1000 rotor
  • Seq:
    04