Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Kaliumkarbonat in aqueous solution USP Test Solution (TS)

Kaliumkarbonat in aqueous solution USP Test Solution (TS)

Plastic bottle, 100 mL
Catalog # 85365.180
Supplier:   VWR Chemicals
Kaliumkarbonat in aqueous solution USP Test Solution (TS)
Catalog # 85365.180
Supplier:  VWR Chemicals
CAS Number:  584-08-7
Kaliumkarbonat in aqueous solution USP Test Solution (TS)
Kaliumkarbonat in aqueous solution USP Test Solution (TS)

Plastic bottle, 100 mL

CAS Number:  584-08-7
Price
262,00 kr
Log in for availability
1 * 100 mL
Product Details & Documents

Ready to use test solution (TS).

  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available
Synonyms:  Potash
  • CAS Number:584-08-7
  • Formula:K₂CO₃
  • MDL Number:MFCD00011382
  • Molecular Weight:138.21 g/mol
  • Merck Index:13,07702
  • Melting Pt:891 °C
  • Storage Temperature:Stue temperatur
  • EINECS:0
Specifications

Specifications

  • Pakning:
    Plastic bottle
  • Pakn:
    100 mL

Specifications