Orders LinkContactUsLinkComponent
VWR®, Density standards referenced at 20 °C
VWR®, Density standards referenced at 20 °C
Density standard, 2,4219 g/ml, 20 °C, 100 mL
Katalog # 85195.180
Leverandør:   VWR Chemicals
VWR®, Density standards referenced at 20 °C
Katalog # 85195.180
Leverandør:  VWR Chemicals
CAS nummer:  
VWR®, Density standards referenced at 20 °C
VWR®, Density standards referenced at 20 °C
Density standard, 2,4219 g/ml, 20 °C, 100 mL
CAS nummer:  
Price
1.970,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents
These products can be used as calibration standards for density measurements by pycnometric, vibrational or hydrometer-based techniques.
  • High accuracy across an extensive density range (0,6619 to 3,1096 g/ml) at 20 °C
  • No toxic heavy metals used in any formulation
  • Can be used with any brand or type of density measuring instrument
  • Supplied in a high quality, tamper-proof, amber glass bottle
These standards are prepared gravimetrically (solute and solvent) using OIML traceable weights. The density values of the standards are established using fully calibrated reference pycnometers.
Certifications: In accordance with ASTM D1480-12 for testing of density or relative density. A certificate of calibration is issued in accordance with the requirements of ISO/IEC 17025.
Specifikationer
Specifikationer
  • Pakn:
    100 mL
  • Referencetemperatur:
    20 °C
  • Beskrivelse:
    Density standard
  • Massefylde:
    2,4219 g/ml
Specifikationer