VWR®, Density standards referenced at 20 °C
Density standard, 1,3304 g/ml, 20 °C, 100 mL
Leverandør: VWR Chemicals
VWR®, Density standards referenced at 20 °C
Katalog # 85188.180
Leverandør: VWR Chemicals
CAS nummer:
Gå til:
Product Details & Documents
These products can be used as calibration standards for density measurements by pycnometric, vibrational or hydrometer-based techniques.
- High accuracy across an extensive density range (0,6619 to 3,1096 g/ml) at 20 °C
- No toxic heavy metals used in any formulation
- Can be used with any brand or type of density measuring instrument
- Supplied in a high quality, tamper-proof, amber glass bottle
These standards are prepared gravimetrically (solute and solvent) using OIML traceable weights. The density values of the standards are established using fully calibrated reference pycnometers.
Certifications: In accordance with ASTM D1480-12 for testing of density or relative density. A certificate of calibration is issued in accordance with the requirements of ISO/IEC 17025.
Documents
Certifikater
Søg efter partinummer23117296.pdf SDS_DA_DK_VWRC84004.pdf SDS_DA_DK_VWRC85170.pdf SDS_DA_DK_VWRC85172.pdf SDS_DA_DK_VWRC85176.pdf
SDS
More
VWR® for Calibrations
Produktlitteratur
Specifikationer
Specifikationer
- Pakn:100 mL
- Referencetemperatur:20 °C
- Beskrivelse:Density standard
- Massefylde:1,3304 g/ml



