VWR®, Density standards referenced at 20 °C
Density standard, 1,2486 g/ml, 20 °C, 100 mL
Leverandør: VWR Chemicals
VWR®, Density standards referenced at 20 °C
Katalog # 85187.180
Leverandør: VWR Chemicals
CAS nummer:
Gå til:
Product Details & Documents
These products can be used as calibration standards for density measurements by pycnometric, vibrational or hydrometer-based techniques.
- High accuracy across an extensive density range (0,6619 to 3,1096 g/ml) at 20 °C
- No toxic heavy metals used in any formulation
- Can be used with any brand or type of density measuring instrument
- Supplied in a high quality, tamper-proof, amber glass bottle
These standards are prepared gravimetrically (solute and solvent) using OIML traceable weights. The density values of the standards are established using fully calibrated reference pycnometers.
Certifications: In accordance with ASTM D1480-12 for testing of density or relative density. A certificate of calibration is issued in accordance with the requirements of ISO/IEC 17025.
Documents
Certifikater
Søg efter partinummer23117296.pdf SDS_DA_DK_VWRC84004.pdf SDS_DA_DK_VWRC85170.pdf SDS_DA_DK_VWRC85172.pdf SDS_DA_DK_VWRC85176.pdf
SDS
More
VWR® for Calibrations
Product Literature
Specifikationer
Specifikationer
- Pakn:100 mL
- Referencetemperatur:20 °C
- Beskrivelse:Density standard
- Massefylde:1,2486 g/ml
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



