Orders LinkContactUsLinkComponent
VWR®, COD calibration standards
VWR®, COD calibration standards
COD standard solution, 1300 mg/L, Plastic bottle, 500 mL
Katalog # 85051.260
Leverandør:   VWR Chemicals
VWR®, COD calibration standards
Katalog # 85051.260
Leverandør:  VWR Chemicals
CAS nummer:  
VWR®, COD calibration standards
VWR®, COD calibration standards
COD standard solution, 1300 mg/L, Plastic bottle, 500 mL
CAS nummer:  
Price
1.160,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents
These standards are ideal to use as control standards for COD measurements to verify that correct analysis for COD has taken place.
Achieving an acceptable result for a control standard will improve confidence in sample readings for this important environmental parameter.
Specifikationer
Specifikationer
  • Koncentration:
    1300 mg/L
  • Pakning:
    Plastic bottle
  • Pakn:
    500 mL
  • Beskrivelse:
    COD standard solution