Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Cleaning brushes for samplers

Cleaning brushes for samplers

Nylon, 7 - 14 mm, Wire, 2100 mm
Katalog # 300-0515
Leverandør:   Sampling Systems
Cleaning brushes for samplers
Katalog # 300-0515
Leverandør:  Sampling Systems
Cleaning brushes for samplers
Cleaning brushes for samplers

Nylon, 7 - 14 mm, Wire, 2100 mm

Price
585,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Brushes with nylon bristles in a twisted stainless steel wire frame and aluminium handle. Ideal for cleaning both stainless steel and plastic samplers.

  • Available in a range of lengths and diameters

Documents

Specifikationer
  • Børstemateriale:
    Nylon
  • Børstehoved Ø:
    7 - 14 mm
  • Handle material:
    Wire
  • Seq:
    04
  • Samlet længde:
    2100 mm