Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Cleaning brushes for samplers

Cleaning brushes for samplers

Nylon, 30 - 45 mm, Wire, 2100 mm
Katalog # 300-0498
Leverandør:   Sampling Systems
Cleaning brushes for samplers
Katalog # 300-0498
Leverandør:  Sampling Systems
Cleaning brushes for samplers
Cleaning brushes for samplers

Nylon, 30 - 45 mm, Wire, 2100 mm

Price
585,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Brushes with nylon bristles in a twisted stainless steel wire frame and aluminium handle. Ideal for cleaning both stainless steel and plastic samplers.

  • Available in a range of lengths and diameters

Documents

Specifikationer
  • Børstemateriale:
    Nylon
  • Børstehoved Ø:
    30 - 45 mm
  • Handle material:
    Wire
  • Seq:
    12
  • Samlet længde:
    2100 mm
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.