Squeegee mop and handles
Cleanroom handle, length 1,45 m
Catalog # PPSA3590200
Supplier: PFENNIG REINIGUNGSTECHNIK
Jump to:
Product Details & Documents
Squeegee mops and handles for hygienically sensitive areas.
- Mop: PP (body), TPE (lip)
- Hygienic squeegee is suitable with cleanroom and telescopical handles
- Handles are anodised aluminum tubes
Specifications
- Beskrivelse:Cleanroom handle, length 1,45 m
- Seq:02