Orders LinkContactUsLinkComponent
VWR®, Viscosity standards
160 cSt, 120 cSt, 60 cSt, 54 cSt, 35 cSt, 500 mL
Katalog # 85098.260
Leverandør:   VWR Chemicals
VWR®, Viscosity standards
Katalog # 85098.260
Leverandør:  VWR Chemicals
CAS nummer:  
VWR®, Viscosity standards
VWR®, Viscosity standards
160 cSt, 120 cSt, 60 cSt, 54 cSt, 35 cSt, 500 mL
CAS nummer:  
Price
739,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents
These standards can be used for calibration, control, verification, qualification or method validation of kinematic and dynamic viscosity measurement instruments (both manual and automatic). Each standard is certified for kinematic viscosity (mm²/s, cSt), dynamic viscosity (cP) and density (g/ml) at a range of temperatures.
  • Manufactured from high quality, stable base oils and additives
  • Extended shelf life with secure packaging
  • All standards observe Newtonian fluid behaviour
These standards are traceable to the ITS 90 temperature scale and the universally accepted primary standard value of the viscosity of water at 20 °C, defined as 1,0034 mm²/s (cSt) by ISO 3666.
 : Certified according to ASTM D2162 using Ubbelohde master viscometers and ISO 17025 accreditation for calibration.
Specifikationer
Specifikationer
  • Pakn:
    500 mL
  • Viscositet ved 20 °C:
    160 cSt
  • Viscositet ved 25 °C:
    120 cSt
  • Viscositet ved 37,78 °C:
    60 cSt
  • Viscositet ved 40 °C:
    54 cSt
  • Viscositet ved 50 °C:
    35 cSt
Specifikationer