Accessories for multi-mode Varioskan™ reader
TRF emission filter, 665 nm HBW10 D25 mm
Catalog # LABLF66510
Supplier: THERMO LABSYSTEMS LIFE SCIENCE
Accessories for multi-mode Varioskan™ reader
Catalog # LABLF66510
Supplier: THERMO LABSYSTEMS LIFE SCIENCE
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:TRF emission filter, 665 nm HBW10 D25 mm
- Seq:23