Accessories for multi-mode Varioskan™ reader
TRF emission filter, 520 nm HBW10 D25 mm
Katalog # LABLF52010
Leverandør: THERMO LABSYSTEMS LIFE SCIENCE
Accessories for multi-mode Varioskan™ reader
Katalog # LABLF52010
Leverandør: THERMO LABSYSTEMS LIFE SCIENCE
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:TRF emission filter, 520 nm HBW10 D25 mm
- Seq:21