VWR®, Carboys, Wide Neck, Rectangular, with Stopcock
HDPE or PP translucent, with purple screw caps.
Use for storing and dispensing solutions and media. Rectangular carboys feature a heavy-duty stainless steel handle for easy carrying and pouring. Carboys are leakproof on standard test conditions. Graduated to identify the volume dispensed. Stopcocks are tied to the handle and require hand assembly for use. No tools are necessary.
HDPE carboys
These high-density polyethylene (HDPE), disposable carboys are ideal for storing and transfer of sterile fluids and pharmaceutical/biotech reagents.
Good for cold storage (down to −80 °C), working temperature of −80 to +120 °C
High impact strength, even at negative temperatures
Widely used for long storage due to high chemical resistance property
Widely used in food and milk industries
PP carboys
Ideal for sterile water storage, polypropylene carboys can be autoclaved before refilling to stop recurring bacterial growth.
Excellent chemical resistance
Working temperature: 0 to 135 °C
Autoclavable
Note: Polypropylene carboys are autoclavable. HDPE carboys are not autoclavable. Always remove stopcock and cap from the carboy before autoclaving.
Certifications: Resin meets US FDA 21 CFR, USP Class VI
Documents
Avantor Cell Culture Solutions.pdf VWR Carboys brochure_Update_MR VWR® Lab bric-a-brac
Product Literature
spec_VWR Carboy
Datasheet
- Gevind:100 mm
- Kapacitet:20 L
- Seq:02
- Hals indv. Ø:83 mm
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



