Reusable half masks, 6500
6502QL, M
Katalog # 111-1664
Leverandør: 3M
Gå til:
Product Details & Documents
Reusable, overmoulded, low profile designed masks offer a wide field of vision. They are compatible with 3M™ welding/grinding shields and low in maintenance with minimal parts and crevices.
- Lightweight silicone faceseal provides comfort and stability with a soft but firm seal keeping its shape in hot environments
- Adjustable head harness assembly with three-size adjustable head cradle and durable straps provides a comfortable fit
- Flexible bayonet system (3M Bayonet Connection™) for connection to a wide range of 3M™ gas, vapour and particulate filters
- 3M Cool Flow Exhalation Valve™ ensures enhanced comfort by reducing heat and moisture and less exhalation resistance
- Valve cover design directs exhaled breath and moisture downward and features an easy positive pressure seal check
- Quick Latch (QL)-Drop Down Mechanis (QL version only): Easy on and off, safety helmets or face shields can be kept on
Certifications: EN 140:1998
Specifikationer
- Model:6502QL
- Seq:04
- Størrelse:M