Orders LinkContactUsLinkComponent
Reusable half masks, 6500
Reusable half masks, 6500
6503QL, L
  111-1666
 :   3M
Reusable half masks, 6500
  111-1666
 :  3M
Reusable half masks, 6500
Reusable half masks, 6500
6503QL, L
359,00 kr
1 * 1 Items
Reusable, overmoulded, low profile designed masks offer a wide field of vision. They are compatible with 3M™ welding/grinding shields and low in maintenance with minimal parts and crevices.
  • Lightweight silicone faceseal provides comfort and stability with a soft but firm seal keeping its shape in hot environments
  • Adjustable head harness assembly with three-size adjustable head cradle and durable straps provides a comfortable fit
  • Flexible bayonet system (3M Bayonet Connection™) for connection to a wide range of 3M™ gas, vapour and particulate filters
  • 3M Cool Flow Exhalation Valve™ ensures enhanced comfort by reducing heat and moisture and less exhalation resistance
  • Valve cover design directs exhaled breath and moisture downward and features an easy positive pressure seal check
  • Quick Latch (QL)-Drop Down Mechanis (QL version only): Easy on and off, safety helmets or face shields can be kept on
 : EN 140:1998
  • Model:
    6503QL
  • Seq:
    06
  • Størrelse:
    L