Overalls, PP, ST11
mørkeblå, 3XL
Katalog # PRTWST11NARXXXL
Leverandør: Portwest
Gå til:
Product Details & Documents
This disposable lightweight overall are made from a breathable non woven PP fabric, enabling the user to keep cool while wearing the garment.
- Generous size design provides exceptional comfort
- Shields against non-hazardous dirt, dust and grime
Weight: 40 g
Specifikationer
- Tøj størrelse:3XL
- Farve:mørkeblå
- Seq:11
- Cat. No.:PRTWST11NARXXXL
- Vægt:40 g/m²
- Model:ST11
- Outfit applikationer:General Purpose|Food