Accessories for Loopster basic and digital
Spare clips with Ø25 mm for use with Universal disc
Catalog # 444-0952
Supplier: IKA
Jump to:
Product Details & Documents
Specifications
- Beskrivelse:Spare clips with Ø25 mm for use with Universal disc
- Seq:12
Accessories
Product Information
- Catalog #BeskrivelseAvailabilityPrice
- 444-0952Spare clips with Ø25 mm for use with Universal discLog in for availability1 * 12 Items - 384,00 kr
Product Information
- Catalog #BeskrivelseAvailabilityPrice
- 444-0857Spare clips with Ø12 mm for use with Universal discLog in for availability1 * 12 Items - 384,00 kr
Product Information
- Catalog #BeskrivelseAvailabilityPrice
- 444-0856Spare clips with Ø10 mm for use with Universal discLog in for availability1 * 12 Items - 384,00 kr
Product Information
- Catalog #BeskrivelseAvailabilityPrice
- 444-0953Spare clips with Ø28 mm for use with Universal discLog in for availability1 * 12 Items - 384,00 kr
Product Information
- Catalog #BeskrivelseAvailabilityPrice
- 444-0951Spare clips with Ø16 mm for use with Universal discLog in for availability1 * 12 Items - 384,00 kr