Objektglas med fordybning
Microscope slides with reaction wells (numbered), grøn, 10, 8 mm
Katalog # MARI1216523
Leverandør: MARIENFELD
Gå til:
Product Details & Documents
Cavity slides for diagnosis with well wettable reaction wells and frosted marking area of 20 mm at one end.
- Soda-lime glass with epoxy resin colour masks
- High resistance against solvents
Specifikationer
- Ant. brønde:10
- Farve:grøn
- Beskrivelse:Microscope slides with reaction wells (numbered)
- Seq:33
- Brønd Ø:8 mm