Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Powder lances

Powder lances

Powder lance, Rustfrit stål 316, 1030 mm, 300 ml
Katalog # SAMP2250A-1000
Leverandør:   Sampling Systems
Powder lances
Katalog # SAMP2250A-1000
Leverandør:  Sampling Systems
Powder lances
Powder lances

Powder lance, Rustfrit stål 316, 1030 mm, 300 ml

Price
3.970,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

The powder lance can be used to manually take large volume samples.

  • Cross sectional sampling without damage
  • Mostly used with cohesive powders
  • Horizontal use through a sack or elsewhere vertically

Suitable for use in the pharmaceutical industries.

Specifikationer
  • Volumen:
    300 ml
  • Beskrivelse:
    Powder lance
  • Længde:
    1030 mm
  • Materiale:
    Rustfrit stål 316
  • Seq:
    02