Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
QuEChERS standards for AOAC method 2007.01

QuEChERS standards for AOAC method 2007.01

QC spike mix, 40 µg/ml in acetonitrile:acetic acid (99.9:0.1), 5 ml/ampoule, 1 Ampoul
Katalog # REST31999
Leverandør:   Restek
QuEChERS standards for AOAC method 2007.01
Katalog # REST31999
Leverandør:  Restek
CAS nummer:  
QuEChERS standards for AOAC method 2007.01
QuEChERS standards for AOAC method 2007.01

QC spike mix, 40 µg/ml in acetonitrile:acetic acid (99.9:0.1), 5 ml/ampoule, 1 Ampoul

CAS nummer:  
Price
2.030,00 kr
Log ind for tilgængelighed
1 * 5 mL
Product Details & Documents

QuEChERS standards are ready to use for generating test mixes, calibration standards, and spiking experiments.

  • Reliable standards produced according to specifications defined in AOAC official method 2007.01
  • Cost-effective QuEChERS standards can be used without dilutions for greater lab efficiency

Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.

These standards includes internal standard mix, triphenylphosphate (TPP) solution, and QC spike mix.

QC spike mix contains 27 components: atrazine (1912-24-9); azoxystrobin (131860-33-8); bifenthrin (82657-04-3); carbaryl (sevin) (63-25-2); chlorothalonil (1897-45-6); chlorpyrifos (2921-88-2); chlorpyrifos methyl (5598-13-0); lambda-cyhalothrin (91465-08-6); cyprodinil (121552-61-2); 2,4'-ddd (53-19-0); dichlorvos (ddvp) (62-73-7); endosulfan sulfate (1031-07-8); ethion (563-12-2); imazalil (35554-44-0); imidacloprid (138261-41-3); kresoxim methyl (143390-89-0); linuron (330-55-2); methamidophos (10265-92-6); methomyl (16752-77-5); cis-permethrin (61949-76-6); trans-permethrin (61949-77-7); procymidone (32809-16-8); pymetrozine (123312-89-0); tebuconazole (107534-96-3); thiabendazole (148-79-8); tolylfluanid (731-27-1); trifluralin (1582-09-8).

Specifikationer

Specifikationer

  • Pakn:
    1 Ampoul
  • Beskrivelse:
    QC spike mix, 40 µg/ml in acetonitrile:acetic acid (99.9:0.1)
  • Pk Info:
    5 ml/ampoule

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.