Revised B/N surrogate mix
Revised B/N Surrogate Mix, 5000 µg/ml, methylene chloride, 5 ml/ampule, 5 mL
REST31889
: Restek
This mix contains four components (2-fluorobiphenyl, nitrobenzene-d5, p-terphenyl-d14, pyrene-d10).
- Pakn:5 mL
- Beskrivelse:Revised B/N Surrogate Mix, 5000 µg/ml, methylene chloride, 5 ml/ampule