Orders LinkContactUsLinkComponent
GC-MS cleaning kit
GC-MS cleaning kit
Mass spec cleaning kit without Dremel tool
Katalog # REST27195
Leverandør:   Restek
GC-MS cleaning kit
Katalog # REST27195
Leverandør:  Restek
GC-MS cleaning kit
GC-MS cleaning kit
Mass spec cleaning kit without Dremel tool
Price
2.430,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
This GC-MS cleaning kit has assembled all of the necessary components for cleaning and polishing your ion source.
Leveringsinformation: Kit includes lint-free nylon gloves (small, 2 pair), lint-free nylon gloves (large, 2 pair), lint-free cotton cloth, 9×9 (10 packs), micro mesh 4×6 sheet (4 packs), aluminum oxide (1-kg jar), cotton tip applicators, large tweezers, small tweezers, septum puller, Dremel® tool, battery-operated and tool kit bag.
Specifikationer
  • Beskrivelse:
    Mass spec cleaning kit without Dremel tool
  • Seq:
    02
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Mass spec cleaning kit replacement parts kit include lint-free nylon gloves (small, 2 pair), lint-free nylon gloves (large, 2 pair), lint-free cotton cloth, 9×9 (10 packs), micro mesh 4×6 sheet (4 packs)
    Mass spec cleaning kit replacement parts kit include lint-free nylon gloves (small, 2 pair), lint-free nylon gloves (large, 2 pair), lint-free cotton cloth, 9×9 (10 packs), micro mesh 4×6 sheet (4 packs)
    REST27196
    Log ind for tilgængelighed
    1 * 1 Items - 784,00 kr