GC-MS cleaning kit
Mass spec cleaning kit without Dremel tool
Katalog # REST27195
Leverandør: Restek
Gå til:
Product Details & Documents
This GC-MS cleaning kit has assembled all of the necessary components for cleaning and polishing your ion source.
Leveringsinformation: Kit includes lint-free nylon gloves (small, 2 pair), lint-free nylon gloves (large, 2 pair), lint-free cotton cloth, 9×9 (10 packs), micro mesh 4×6 sheet (4 packs), aluminum oxide (1-kg jar), cotton tip applicators, large tweezers, small tweezers, septum puller, Dremel® tool, battery-operated and tool kit bag.
Specifikationer
- Beskrivelse:Mass spec cleaning kit without Dremel tool
- Seq:02
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Mass spec cleaning kit replacement parts kit include lint-free nylon gloves (small, 2 pair), lint-free nylon gloves (large, 2 pair), lint-free cotton cloth, 9×9 (10 packs), micro mesh 4×6 sheet (4 packs)Mass spec cleaning kit replacement parts kit include lint-free nylon gloves (small, 2 pair), lint-free nylon gloves (large, 2 pair), lint-free cotton cloth, 9×9 (10 packs), micro mesh 4×6 sheet (4 packs)REST27196Log ind for tilgængelighed1 * 1 Items - 784,00 kr