Orders LinkContactUsLinkComponent
Waste overflow indicator for HPLC systems
Waste overflow indicator for HPLC systems
Waste overflow indicator for HPLC systems, 4 L bottles
Katalog # REST26543
Leverandør:   Restek
Waste overflow indicator for HPLC systems
Katalog # REST26543
Leverandør:  Restek
Waste overflow indicator for HPLC systems
Waste overflow indicator for HPLC systems
Waste overflow indicator for HPLC systems, 4 L bottles
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
The waste overflow indicator keeps the mobile phase waste in the waste container.
  • Avoid messy pooling around mobile phase waste containers
  • Audible alarm instantly alerts user, preventing overflow
  • Compact, battery operated unit
Compact, battery operated unit fits securely on solvent bottles and accommodates two waste streams. An audible alarm is given as the solvent waste container approaches capacity, giving you time to empty or change the container.
Specifikationer
  • Til:
    4 L bottles
  • Beskrivelse:
    Waste overflow indicator for HPLC systems
  • Seq:
    01
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Replacement AA batteries (3×) for waste overflow indicator
    Replacement AA batteries (3×) for waste overflow indicator
    REST26545
    Log ind for tilgængelighed
    1 * 3 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Replacement AA battery (1×) for waste overflow indicator
    Replacement AA battery (1×) for waste overflow indicator
    REST26544