Waste overflow indicator for HPLC systems
Waste overflow indicator for HPLC systems, 4 L bottles
Katalog # REST26543
Leverandør: Restek
Gå til:
Product Details & Documents
The waste overflow indicator keeps the mobile phase waste in the waste container.
- Avoid messy pooling around mobile phase waste containers
- Audible alarm instantly alerts user, preventing overflow
- Compact, battery operated unit
Compact, battery operated unit fits securely on solvent bottles and accommodates two waste streams. An audible alarm is given as the solvent waste container approaches capacity, giving you time to empty or change the container.
Specifikationer
- Til:4 L bottles
- Beskrivelse:Waste overflow indicator for HPLC systems
- Seq:01
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Replacement AA batteries (3×) for waste overflow indicatorReplacement AA batteries (3×) for waste overflow indicatorREST26545Log ind for tilgængelighed1 * 3 Items
Produktinformation
- Katalog #TilgængelighedPrice
- Replacement AA battery (1×) for waste overflow indicatorReplacement AA battery (1×) for waste overflow indicatorREST26544Log ind for tilgængelighed1 * 1 Items