Orders LinkContactUsLinkComponent
Valves for HPLC
Valves for HPLC
Prime/purge valve for HPLC, 1/4 - 28 flanged seat, stainless steel, with tubing and fittings
Catalog # REST26406
Supplier:   Restek
Valves for HPLC
Catalog # REST26406
Supplier:  Restek
Valves for HPLC
Valves for HPLC
Prime/purge valve for HPLC, 1/4 - 28 flanged seat, stainless steel, with tubing and fittings
Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents
These valves have low internal volumes and are rated to 15000 psi. They feature a two-piece stem assembly in which the rotating upper shank is coupled through a ball joint to a static lower stem.
  • Only fingertight torque is required to make the seal
Specifications
  • Beskrivelse:
    Prime/purge valve for HPLC, 1/4 - 28 flanged seat, stainless steel, with tubing and fittings
  • Seq:
    06