Valves for HPLC
Prime/purge valve for HPLC, 1/4 - 28 flanged seat, stainless steel, with tubing and fittings
Catalog # REST26406
Supplier: Restek
Jump to:
Product Details & Documents
These valves have low internal volumes and are rated to 15000 psi. They feature a two-piece stem assembly in which the rotating upper shank is coupled through a ball joint to a static lower stem.
- Only fingertight torque is required to make the seal
Specifications
- Beskrivelse:Prime/purge valve for HPLC, 1/4 - 28 flanged seat, stainless steel, with tubing and fittings
- Seq:06