Orders LinkContactUsLinkComponent
Valves for HPLC
Valves for HPLC
Side vent valve for HPLC, 1/16" fittings, 1/4 - 28, stainless steel, includes nuts and ferrules
Katalog # REST26403
Leverandør:   Restek
Valves for HPLC
Katalog # REST26403
Leverandør:  Restek
Valves for HPLC
Valves for HPLC
Side vent valve for HPLC, 1/16" fittings, 1/4 - 28, stainless steel, includes nuts and ferrules
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These valves have low internal volumes and are rated to 15000 psi. They feature a two-piece stem assembly in which the rotating upper shank is coupled through a ball joint to a static lower stem.
  • Only fingertight torque is required to make the seal
Specifikationer
  • Beskrivelse:
    Side vent valve for HPLC, 1/16" fittings, 1/4 - 28, stainless steel, includes nuts and ferrules
  • Seq:
    04