Valves for HPLC
Angle valve for HPLC, 1/16" fittings, 1/4 - 28, stainless steel, includes nuts and ferrules
Katalog # REST26401
Leverandør: Restek
Gå til:
Product Details & Documents
These valves have low internal volumes and are rated to 15000 psi. They feature a two-piece stem assembly in which the rotating upper shank is coupled through a ball joint to a static lower stem.
- Only fingertight torque is required to make the seal
Specifikationer
- Beskrivelse:Angle valve for HPLC, 1/16" fittings, 1/4 - 28, stainless steel, includes nuts and ferrules
- Seq:02