Packed GC columns, 2abc refinery column set
Analytical controls configuration
Katalog # REST88000-810
Leverandør: Restek
Gå til:
Product Details & Documents
Special columns for unsaturated light hydrocarbons.
- Faster separations of C1 to C4 hydrocarbons
When used in the proper valving system, it will elute C5+ hydrocarbons ahead of C1 through C4 hydrocarbons.
Specifikationer
- Beskrivelse:Analytical controls configuration
- Seq:01